Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mbnl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134163.4 | NP_598924.1 | 4 | 575 | ||
GenBank mRNA | AB077700.1 | - | 3 | 481 | ||
AF466292.1 | - | 3 | 481 | |||
AJ427920.1 | - | 3 | 481 | |||
AK040522.1 | - | 3 | 329 | |||
AK050908.1 | - | 3 | 294 | |||
AK088971.1 | - | 4 | 588 | |||
AK167342.1 | - | 3 | 294 | |||
BC057157.1 | - | 4 | 813 | 5348959 |