Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Timd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161355.1 | NP_001154827.1 | 8, 9 | 1102 | ||
NM_001161356.1 | NP_001154828.1 | 8, 9 | 875 | |||
NM_134249.5 | NP_599010.4 | 8, 9 | 984 | |||
GenBank mRNA | AF399828.1 | - | 8, 9 | 836 | ||
AK145630.1 | - | 8, 9 | 863 | |||
AK163225.1 | - | 8, 9 | 863 | |||
AK166877.1 | - | 8, 9 | 856 | |||
BC025096.1 | - | 8, 9 | 914 | 4158605 | ||
BC028829.1 | - | 8, 9 | 946 | 5102129 |