Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il17rd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134437.3 | NP_602319.1 | 12 | 1791 | ||
GenBank mRNA | AF424804.1 | - | 8 | 1220 | ||
AF459444.1 | - | 12 | 1710 | |||
AF494209.1 | - | 8 | 1500 | |||
AF494210.1 | - | 12 | 1851 | |||
AK133233.1 | - | 14 | 1954 | |||
AK139904.1 | - | 12 | 1718 | |||
BC138629.1 | - | 12 | 1746 | 8861649 | ||
CD564540.1 | - | 1 | 299 |