Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpr37l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB016602.1 | - | 2 | 1925 | ||
AK158275.1 | - | 2 | 2185 | |||
BC098482.1 | - | 2 | 1873 | 30103328 |