Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Triobp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024716.1 | NP_001019887.1 | 6 | 956 | ||
NM_001039156.1 | NP_001034245.1 | 17 | 5723 | |||
NM_138579.4 | NP_613045.3 | 16 | 5585 | |||
GenBank mRNA | AK053194.1 | - | 6 | 916 | ||
AK085512.1 | - | 6 | 927 | |||
AK129417.1 | - | 5 | 2381 | |||
AK155197.1 | - | 9 | 3288 | |||
AK170390.1 | - | 6 | 902 | |||
AK178072.1 | - | 1 | 21 | |||
AK186966.1 | - | 1 | 21 | |||
BC003984.1 | - | 4 | 752 | |||
BC042760.1 | - | 6 | 895 | |||
BC053329.1 | - | 6 | 940 | |||
CA751843.1 | - | 1 | 402 | |||
DQ228000.1 | - | 17 | 5723 | |||
DQ228001.2 | - | 16 | 5585 | |||
DQ278602.1 | - | 15 | 5204 |