Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005967.3 | NP_005958.1 | 3 | 1381 | ||
XM_005268894.2 | XP_005268951.1 | 3 | 1381 | |||
GenBank mRNA | AK314229.1 | - | 3 | 1161 | ||
BC065931.1 | - | 3 | 1168 | 6157017 | ||
U48361.1 | - | 3 | 1160 |