Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dgkg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138650.2 | NP_619591.1 | 25 | 2817 | ||
GenBank mRNA | AK083258.1 | - | 24 | 2693 | ||
AK220294.1 | - | 25 | 2759 | |||
BC015278.1 | - | 24 | 2390 | 4221290 |