Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bspry | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138653.1 | NP_619594.1 | 6, 7 | 1434 | ||
GenBank mRNA | AJ276690.1 | - | 6, 7 | 1434 | ||
BC139079.1 | - | 6, 7 | 1450 | 8862101 | ||
BC139080.1 | - | 6, 7 | 1450 | 8862102 | ||
BC145535.1 | - | 7, 8 | 1075 | 9054098 |