Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRCC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005973.4 | NP_005964.3 | 7 | 2040 | ||
XM_005245313.2 | XP_005245370.1 | 8 | 2136 | |||
XM_005245314.1 | XP_005245371.1 | 6 | 1944 | |||
GenBank mRNA | AK126403.1 | - | 2 | 1891 | ||
BC004913.2 | - | 7 | 1956 | 3535413 | ||
BC010450.1 | - | 7 | 1968 | 4179105 | ||
BU942864.1 | - | 4 | 646 | |||
X97124.1 | - | 7 | 1940 |