Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: S100A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005978.3 | NP_005969.1 | 3 | 682 | ||
GenBank mRNA | AF086003.1 | - | 2 | 337 | ||
BC002829.2 | - | 3 | 682 | 3659591 | ||
BC105787.1 | - | 3 | 386 | 6702470 | ||
BQ953250.1 | - | 377 | ||||
M87068.1 | - | 3 | 384 |