Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Shkbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138676.2 | NP_619617.2 | 8 | 666 | ||
GenBank mRNA | AF246218.1 | - | 8 | 672 | ||
AK047434.1 | - | 6 | 797 | |||
AK154779.1 | - | 8 | 655 | |||
AK160641.1 | - | 7 | 738 | |||
AK190932.1 | - | 2 | 191 | |||
BC061477.1 | - | 8 | 619 | 6336438 |