Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Shkbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138676.2 | NP_619617.2 | 11 | 1035 | ||
GenBank mRNA | AF246218.1 | - | 11 | 1041 | ||
AK047434.1 | - | 9 | 1166 | |||
AK154779.1 | - | 11 | 1023 | |||
AK160641.1 | - | 10 | 1107 | |||
AK198381.1 | - | 1 | 29 | |||
BC061477.1 | - | 11 | 988 | 6336438 |