Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Shkbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138676.2 | NP_619617.2 | 14 | 1413 | ||
GenBank mRNA | AF246218.1 | - | 14 | 1419 | ||
AK047434.1 | - | 12 | 1544 | |||
AK047826.1 | - | 2 | 212 | |||
AK154779.1 | - | 14 | 1401 | |||
AK160641.1 | - | 13 | 1485 | |||
BC061477.1 | - | 14 | 1366 | 6336438 |