Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138756.4 | NP_620095.1 | 7 | 1185 | ||
GenBank mRNA | AK034423.1 | - | 3 | 721 | ||
AK042075.1 | - | 7 | 1274 | |||
AK053646.1 | - | 7 | 1301 | |||
AK078779.1 | - | 7 | 1186 | |||
BC004098.1 | - | 1 | 378 | |||
BC008171.1 | - | 7 | 1177 | 3710468 |