Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138756.4 | NP_620095.1 | 6, 7 | 839 | ||
GenBank mRNA | AK034423.1 | - | 2, 3 | 375 | ||
AK042075.1 | - | 6, 7 | 928 | |||
AK053646.1 | - | 6, 7 | 955 | |||
AK078779.1 | - | 6, 7 | 840 | |||
BC004098.1 | - | 32 | ||||
BC008171.1 | - | 6, 7 | 831 | 3710468 |