Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ripk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138952.3 | NP_620402.1 | 11 | 1401 | ||
GenBank mRNA | AF461040.1 | - | 11 | 1299 | ||
AF487539.1 | - | 11 | 1299 | |||
AK159593.1 | - | 11 | 1425 | |||
AK172319.1 | - | 11 | 1423 | |||
AK172417.1 | - | 11 | 1423 | |||
BC025611.1 | - | 9 | 919 | |||
BC050845.1 | - | 11 | 1415 | |||
BC058088.1 | - | 12 | 1670 | |||
BC069878.1 | - | 11 | 1426 | 5316066 |