Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rfpl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145013.1 | NP_001138485.1 | 2 | 323 | ||
NM_138954.3 | NP_620404.1 | 2 | 555 | |||
GenBank mRNA | AK136022.1 | - | 2 | 324 | ||
AK139569.1 | - | 2 | 557 | |||
AY070253.1 | - | 2 | 545 | |||
BC069834.1 | - | 2 | 309 | 30474534 |