Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ogt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290535.1 | NP_001277464.1 | 2 | 199 | ||
NM_139144.4 | NP_631883.2 | 2 | 396 | |||
GenBank mRNA | AF363030.1 | - | 2 | 161 | ||
AK076418.1 | - | 3 | 279 | |||
AK149194.1 | - | 1 | 25 | |||
AK163296.1 | - | 2 | 356 | |||
BC025097.1 | - | 2 | 339 | |||
BC037194.1 | - | 2 | 280 | |||
BC055851.1 | - | 2 | 251 | |||
BC057319.1 | - | 2 | 319 | 6830845 | ||
BY196484.1 | - | 3 | 290 | |||
CB236717.1 | - | 2 | 386 | |||
CB520299.1 | - | 2 | 146 | |||
CD544482.1 | - | 3 | 262 | |||
CF166269.1 | - | 2 | 374 |