Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ogt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290535.1 | NP_001277464.1 | 3 | 317 | ||
NM_139144.4 | NP_631883.2 | 3 | 514 | |||
GenBank mRNA | AF363030.1 | - | 3 | 279 | ||
AK076418.1 | - | 4 | 397 | |||
AK149194.1 | - | 2 | 143 | |||
AK163296.1 | - | 3 | 474 | |||
BC025097.1 | - | 3 | 457 | |||
BC037194.1 | - | 3 | 398 | |||
BC055851.1 | - | 3 | 369 | |||
BC057319.1 | - | 3 | 437 | 6830845 | ||
CB236717.1 | - | 3 | 504 | |||
CB520299.1 | - | 3 | 264 | |||
CD544482.1 | - | 4 | 380 | |||
CF166269.1 | - | 3 | 492 |