Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arap3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205336.1 | NP_001192265.1 | 5, 6 | 1147 | ||
NM_139206.2 | NP_631945.2 | 5, 6 | 947 | |||
GenBank mRNA | AF469621.1 | - | 4, 5 | 897 | ||
AF469622.1 | - | 5, 6 | 1088 | |||
AK036638.1 | - | 5, 6 | 912 | |||
AK053122.1 | - | 4, 5 | 1337 | |||
AK220486.1 | - | 5, 6 | 940 | |||
BC068145.1 | - | 5, 6 | 935 | 6856630 |