Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Galnt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139272.2 | NP_644678.2 | 16 | 3304 | ||
GenBank mRNA | AK145122.1 | - | 16 | 3179 | ||
AK149609.1 | - | 16 | 3234 | |||
AK151536.1 | - | 16 | 3096 | |||
AK185552.1 | - | 1 | 71 | |||
AK200523.1 | - | 1 | 154 | |||
BC007172.1 | - | 15 | 3040 | |||
BC025877.1 | - | 1 | 287 | |||
BC039641.1 | - | 1 | 249 | |||
BC053063.1 | - | 15 | 3148 | 5702572 | ||
BC059818.1 | - | 16 | 3248 | 5697789 |