Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ece2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139293.2 | NP_647454.2 | 4 | 556 | ||
NM_177940.1 | NP_808809.1 | 4 | 776 | |||
NM_177941.1 | NP_808810.1 | 5 | 863 | |||
NM_177942.1 | NP_808811.1 | 3 | 469 | |||
GenBank mRNA | AF396699.1 | - | 4 | 497 | ||
AF489569.1 | - | 4 | 595 | |||
AF489570.1 | - | 5 | 682 | |||
AF489571.1 | - | 3 | 469 | |||
AF489572.1 | - | 4 | 556 | |||
AK031951.1 | - | 4 | 564 | |||
AK043473.1 | - | 2 | 219 | |||
AU080537.1 | - | 4 | 513 | |||
BC138052.1 | - | 4 | 463 | 8861071 | ||
BC138053.1 | - | 4 | 463 | 8861072 | ||
BC145126.1 | - | 4 | 463 |