Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mylk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139300.3 | NP_647461.3 | 16 | 3053 | ||
GenBank mRNA | AK078556.1 | - | 1 | 492 | ||
AY237726.1 | - | 16 | 3082 | |||
AY237727.2 | - | 1 | 606 | |||
BC030478.1 | - | 6 | 1285 | |||
BC045197.1 | - | 1 | 536 | |||
BC058610.1 | - | 16 | 3065 | 30056142 | ||
BC145279.1 | - | 16 | 3040 | 9053833 | ||
BC150749.1 | - | 16 | 3040 | 9087660 | ||
BC171954.1 | - | 16 | 3041 |