Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Amhr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144547.2 | NP_653130.2 | 7, 8 | 1066 | ||
GenBank mRNA | AF503863.1 | - | 7, 8 | 1013 | ||
BC119571.1 | - | 7, 8 | 997 | 40057985 |