Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc30a6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252478.1 | NP_001239407.1 | 3, 4 | 236 | ||
NM_144798.6 | NP_659047.2 | 3, 4 | 236 | |||
GenBank mRNA | AF233346.1 | - | 2, 3 | 188 | ||
AF395840.1 | - | 2, 3 | 178 | |||
AK082907.1 | - | 3, 4 | 213 | |||
AK150342.1 | - | 3, 4 | 212 | |||
AK151120.1 | - | 3, 4 | 212 | |||
AK160903.1 | - | 3, 4 | 225 | |||
BC066162.1 | - | 3, 4 | 203 | 30471027 | ||
BC068169.1 | - | 3, 4 | 225 | 30542277 |