Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172100.1 | NP_001165571.1 | 3 | 307 | ||
NM_001172101.1 | NP_001165572.1 | 3 | 310 | |||
NM_145135.3 | NP_660117.2 | 3 | 458 | |||
GenBank mRNA | AF071546.1 | - | 2 | 178 | ||
AK133822.1 | - | 3 | 238 | |||
AK141351.1 | - | 3 | 283 | |||
AK145173.1 | - | 3 | 292 | |||
AK147059.1 | - | 3 | 244 | |||
AK188203.1 | - | 64 | ||||
BC010331.1 | - | 3 | 327 | 2654341 | ||
BM946973.1 | - | 3 | 458 | |||
BY085526.1 | - | 3 | 312 | |||
BY114445.1 | - | 3 | 307 |