Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aif1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145144.1 | NP_660126.1 | 6 | 1595 | ||
GenBank mRNA | AK028955.1 | - | 6 | 1553 | ||
AK045539.1 | - | 6 | 1625 | |||
BC024599.1 | - | 6 | 1595 | 5044495 |