Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf185 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290472.1 | NP_001277401.1 | 7 | 2821 | ||
NM_001290473.1 | NP_001277402.1 | 6 | 2802 | |||
NM_145355.5 | NP_663330.2 | 8 | 2940 | |||
GenBank mRNA | AK034108.1 | - | 8 | 2310 | ||
AK052436.1 | - | 7 | 2230 | |||
AK053358.1 | - | 7 | 2187 | |||
AK077638.1 | - | 7 | 2152 | |||
AK149640.1 | - | 7 | 2172 | |||
AK165895.1 | - | 2243 | ||||
AK167455.1 | - | 7 | 2138 | |||
AK168904.1 | - | 7 | 2175 | |||
BC014812.1 | - | 7 | 2151 | 3153870 |