Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD164 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142401.1 | NP_001135873.1 | 5 | 1931 | ||
NM_001142402.1 | NP_001135874.1 | 5 | 1913 | |||
NM_001142403.1 | NP_001135875.1 | 7 | 1374 | |||
NM_001142404.1 | NP_001135876.1 | 6 | 1278 | |||
NM_006016.4 | NP_006007.2 | 6 | 1970 | |||
GenBank mRNA | AF299341.1 | - | 6 | 1789 | ||
AF299342.1 | - | 5 | 1810 | |||
AF299343.1 | - | 5 | 1828 | |||
AK301692.1 | - | 6 | 1008 | |||
AK307193.1 | - | 3 | 797 | |||
BC011522.2 | - | 6 | 1870 | 3854912 | ||
BC037914.1 | - | 5 | 2472 | |||
BC040317.1 | - | 2 | 3822 | |||
D14043.1 | - | 7 | 1272 | |||
EB388947.1 | - | 1 | 466 |