Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ube4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145400.3 | NP_663375.3 | 17 | 2843 | ||
GenBank mRNA | AK028721.1 | - | 17 | 2628 | ||
AK037131.1 | - | 1 | 125 | |||
AK172893.1 | - | 17 | 2803 | |||
BC006649.1 | - | 19 | 3096 | 3498834 |