Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Edem2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145537.2 | NP_663512.2 | 9 | 1150 | ||
GenBank mRNA | AK029164.1 | - | 9 | 1102 | ||
AK075650.1 | - | 9 | 1064 | |||
AK079296.1 | - | 9 | 1086 | |||
AK080636.1 | - | 9 | 1095 | |||
AK149892.1 | - | 9 | 1085 | |||
AK155024.1 | - | 9 | 1113 | |||
AK166998.1 | - | 9 | 1086 | |||
BC008268.1 | - | 9 | 1061 | 2811074 |