Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rap1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145541.5 | NP_663516.1 | 8 | 1364 | ||
GenBank mRNA | AK030897.1 | - | 8 | 1313 | ||
AK152504.1 | - | 8 | 1284 | |||
BC051419.1 | - | 8 | 1320 | 3494423 | ||
BC083128.1 | - | 7 | 1216 | 6438722 |