Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pttg1ip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145925.2 | NP_666037.1 | 6 | 570 | ||
GenBank mRNA | AK035696.1 | - | 6 | 585 | ||
AK075773.1 | - | 6 | 543 | |||
AK088813.1 | - | 6 | 607 | |||
AK089887.1 | - | 6 | 552 | |||
AK098093.1 | - | 6 | 584 | |||
AK151639.1 | - | 6 | 552 | |||
AK152366.1 | - | 6 | 552 | |||
AK159987.1 | - | 6 | 552 | |||
AK167972.1 | - | 6 | 582 | |||
AK168164.1 | - | 6 | 582 | |||
AK170802.1 | - | 6 | 552 | |||
AK170971.1 | - | 6 | 585 | |||
AK170972.1 | - | 6 | 552 | |||
AK171140.1 | - | 6 | 551 | |||
AK178924.1 | - | 2 | 70 | |||
AK181674.1 | - | 2 | 70 | |||
AK193277.1 | - | 2 | 70 | |||
AK193680.1 | - | 2 | 70 | |||
AK196421.1 | - | 2 | 70 | |||
AK196698.1 | - | 2 | 70 | |||
AK199195.1 | - | 2 | 70 | |||
AK205669.1 | - | 2 | 70 | |||
AK216713.1 | - | 2 | 70 | |||
BC023961.1 | - | 6 | 625 | 5323397 | ||
BC025533.1 | - | 6 | 594 | 5250502 | ||
BC029144.1 | - | 6 | 566 | 4945974 | ||
BC032184.1 | - | 6 | 589 | 5310191 |