Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pttg1ip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145925.2 | NP_666037.1 | 6 | 1119 | ||
GenBank mRNA | AK035696.1 | - | 6 | 1134 | ||
AK075773.1 | - | 6 | 1092 | |||
AK081798.1 | - | 1 | 542 | |||
AK088813.1 | - | 6 | 1156 | |||
AK089887.1 | - | 6 | 1101 | |||
AK098093.1 | - | 6 | 1133 | |||
AK151639.1 | - | 6 | 1101 | |||
AK152366.1 | - | 6 | 1101 | |||
AK159987.1 | - | 6 | 1101 | |||
AK167972.1 | - | 6 | 1130 | |||
AK168164.1 | - | 6 | 1130 | |||
AK170802.1 | - | 6 | 1101 | |||
AK170971.1 | - | 6 | 1134 | |||
AK170972.1 | - | 6 | 1101 | |||
AK171140.1 | - | 6 | 1100 | |||
AK190248.1 | - | 1 | 327 | |||
AK198698.1 | - | 1 | 355 | |||
BC023961.1 | - | 6 | 1174 | 5323397 | ||
BC025533.1 | - | 6 | 1143 | 5250502 | ||
BC029144.1 | - | 6 | 1115 | 4945974 | ||
BC032184.1 | - | 6 | 1138 | 5310191 |