Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fntb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145927.2 | NP_666039.1 | 1 | 113 | ||
GenBank mRNA | AK008228.1 | - | 1 | 148 | ||
AK089911.1 | - | 1 | 143 | |||
BC031417.1 | - | 1 | 113 | 3154478 |