Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fntb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145927.2 | NP_666039.1 | 11 | 1121 | ||
GenBank mRNA | AK089911.1 | - | 11 | 1150 | ||
BC031417.1 | - | 11 | 1121 | 3154478 |