Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Enox2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271447.1 | NP_001258376.1 | 9 | 1304 | ||
NM_001271448.1 | NP_001258377.1 | 8 | 1149 | |||
NM_001271449.1 | NP_001258378.1 | 9 | 1293 | |||
NM_001271450.1 | NP_001258379.1 | 9 | 1293 | |||
NM_001271451.1 | NP_001258380.1 | 9 | 1293 | |||
NM_145951.5 | NP_666063.1 | 9 | 1248 | |||
GenBank mRNA | AK028343.1 | - | 9 | 1294 | ||
AK034058.1 | - | 9 | 1249 | |||
AK041822.1 | - | 9 | 1294 | |||
AK133956.1 | - | 8 | 1120 | |||
AK137736.1 | - | 9 | 1275 | |||
AK139897.1 | - | 9 | 1275 | |||
BC025915.1 | - | 9 | 1267 | 5043650 |