Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbc1d12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145952.3 | NP_666064.3 | 13 | 2229 | ||
GenBank mRNA | AK172979.1 | - | 13 | 2223 | ||
BC033574.1 | - | 4 | 531 | |||
BC138056.1 | - | 13 | 2125 | 8861075 | ||
BC138057.1 | - | 13 | 2125 | 8861076 | ||
BC145129.1 | - | 11 | 1900 | 9053677 | ||
BU152220.1 | - | 2 | 306 |