Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145979.2 | NP_666091.1 | 26 | 4062 | ||
GenBank mRNA | AK147695.1 | - | 26 | 4206 | ||
AK220473.1 | - | 25 | 3970 | |||
BC006035.1 | - | 5 | 627 | |||
BC058578.1 | - | 26 | 4062 | 6489649 | ||
BC059082.1 | - | 13 | 1842 |