Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nrbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147201.2 | NP_671734.1 | 4, 5 | 541 | ||
GenBank mRNA | AF302138.1 | - | 4, 5 | 540 | ||
AK046142.1 | - | 4, 5 | 535 | |||
AK077440.1 | - | 4, 5 | 577 | |||
AK170501.1 | - | 4, 5 | 557 | |||
AK178908.1 | - | 4, 5 | 382 | |||
AK218775.1 | - | 1, 2 | 80 | |||
BC004756.1 | - | 4, 5 | 511 | 3154089 | ||
BC018463.1 | - | 4, 5 | 515 | 3598438 |