Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rps6ka3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_148945.2 | NP_683747.1 | 18 | 1970 | ||
GenBank mRNA | AK154778.1 | - | 18 | 1882 | ||
AY083469.1 | - | 18 | 1632 | |||
BC038683.1 | - | 2 | 56 | |||
BC150156.1 | - | 18 | 1635 | |||
BC150478.1 | - | 18 | 1635 |