Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pknox2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029838.2 | NP_001025009.1 | 10 | 1186 | ||
NM_148950.3 | NP_683752.2 | 9 | 1081 | |||
GenBank mRNA | AB086817.1 | - | 7 | 907 | ||
AF487460.1 | - | 8 | 987 | |||
AK082952.1 | - | 10 | 1131 | |||
BC050865.1 | - | 9 | 1015 | 6332968 |