Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnq3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152923.2 | NP_690887.2 | 6 | 1015 | ||
GenBank mRNA | AY118171.1 | - | 6 | 1015 | ||
BC116306.1 | - | 6 | 1006 | 40095195 |