Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sf3b4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK047751.1 | - | 3 | 374 | ||
AK135543.1 | - | 3 | 382 | |||
AK156828.1 | - | 3 | 379 | |||
AK180912.1 | - | 2 | 271 | |||
BC024418.1 | - | 3 | 417 | 4982610 | ||
BC026567.1 | - | 3 | 405 | 4982239 | ||
BC085273.1 | - | 3 | 357 | 30144329 |