Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc18a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153054.2 | NP_694694.1 | 17 | 2500 | ||
GenBank mRNA | AK076433.1 | - | 16 | 2225 | ||
BC027192.1 | - | 17 | 2438 | 4239930 | ||
BC031146.1 | - | 17 | 2468 | 4974776 |