Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sec63 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153055.3 | NP_694695.3 | 2 | 478 | ||
GenBank mRNA | AK036629.1 | - | 2 | 173 | ||
AK049737.1 | - | 2 | 169 | |||
AY024346.1 | - | 2 | 275 | |||
AY238936.1 | - | 2 | 248 | |||
BC019366.1 | - | 2 | 242 | 5066908 | ||
BC031846.1 | - | 2 | 242 | 5066822 | ||
BC059816.1 | - | 2 | 478 | |||
BC080810.1 | - | 2 | 359 |