Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mmrn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153127.3 | NP_694767.3 | 6 | 1115 | ||
GenBank mRNA | AK077884.1 | - | 6 | 1117 | ||
AK078470.1 | - | 6 | 1091 | |||
AK134285.1 | - | 6 | 1154 | |||
BC145845.1 | - | 6 | 1035 | 40131148 |