Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Unc5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153131.3 | NP_694771.1 | 14, 15 | 2590 | ||
GenBank mRNA | AJ487852.1 | - | 14, 15 | 2591 | ||
AK122575.1 | - | 10, 11 | 2912 | |||
BC058084.1 | - | 13, 14 | 2443 | 6813463 |