Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Unc5d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153135.3 | NP_694775.1 | 5 | 932 | ||
GenBank mRNA | AJ487854.1 | - | 5 | 933 | ||
BC137615.1 | - | 5 | 723 | 8860634 | ||
BC144783.1 | - | 5 | 723 | 9053331 | ||
BC144784.1 | - | 5 | 723 | 9053332 | ||
BC145695.1 | - | 5 | 723 | 40130998 | ||
FJ824810.1 | - | 5 | 865 |