Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dock3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153413.2 | NP_700462.2 | 3 | 542 | ||
GenBank mRNA | AK172930.1 | - | 3 | 501 | ||
AY145302.1 | - | 3 | 492 | |||
BF465833.1 | - | 3 | 216 |